Order Entry
Belgium
ContactUsLinkComponent
Anion exchange chromatography columns, HiTrap Q HP
Anion exchange chromatography columns, HiTrap Q HP
Supplier:  Cytiva
Certificates
undefined
Anion exchange chromatography columns, HiTrap Q HP
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 29-0513-25
  • 17-1153-01
  • 17-1154-01
  • Description:
  • HiTrap™ Q HP
  • HiTrap™ Q HP
  • HiTrap™ Q HP
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 2 - 12
  • 2 - 12
  • 2 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together