Order Entry
Belgium
ContactUsLinkComponent
Short-term storage solutions, 2-8CELLsius
Short-term storage solutions, 2-8CELLsius
Certificates
undefined
Short-term storage solutions, 2-8CELLsius

Specifications

  • Shelf life:
    20 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X
  • Storage temperature:
    2...8 °C

Specifications

Frequently Bought Together