Order Entry
Belgium
ContactUsLinkComponent
 
Cryopreservation solutions, DMSO-free, pZerve™
 
undefined
Cryopreservation solutions, DMSO-free, pZerve™

 

  • Species:
    Human
  • Shelf life:
    18 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X
  • Storage temperature:
    2...8 °C

 

Frequently Bought Together