Order Entry
Belgium
ContactUsLinkComponent
Cryopreservation solution, DMSO-free, Cellvation™
Cryopreservation solution, DMSO-free, Cellvation™
Catalog # PRPH5001
CAS Number:  
undefined
Cryopreservation solution, DMSO-free, Cellvation™
Catalog # PRPH5001
CAS Number:  

Specifications

  • Pk:
    60 mL
  • Species:
    Bovine
  • Description:
    Cryopreservation solution, DMSO-free, Cellvation™
  • Shelf life:
    18 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X
  • Storage temperature:
    2...8 °C

Specifications

About this item

A unique cryopreservation solution that does not contain DMSO or foetal bovine serum. Cellvation™ is convenient to use and provides optimal recovery. Cells can be exposed to Cellvation™ for longer periods of time before the freezing process is initiated. Cellvation™ is ready to use and can be used for cells cultured in serum-free medium or with serum-containing medium.

  • Does not contain DMSO
  • Sterile/ready to use
Certifications: cGMP manufactured.