Serum-free growth medium, defined, HemaPro
Supplier: PROTIDE PHARMACEUTICALS
Certificates
About this item
Serum supplement for the culturing of human progenitor cells. Effective in studying cancer stem cells, iPSC, and mesenchymal stem cells.
- Defined, serum-free, xeno-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilised with a wide variety of basal media
Certifications: cGMP manufactured.
Specifications
- Species:Human
- Total protein:488 µg/ml at 1X
- Shelf life:18 months from DOM
- Endotoxin:<0,5 EU/ml at 1X
- Storage temperature:2...8 °C
Specifications
Frequently Bought Together