Order Entry
Belgium
ContactUsLinkComponent
 
TM-235 defined serum-free growth medium
 
undefined
TM-235 defined serum-free growth medium

 

  • Species:
    Bovine
  • Total protein:
    770 µg/ml at 1X
  • Shelf life:
    18 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X
  • Storage temperature:
    2...8 °C

 

Frequently Bought Together