ContactUsLinkComponent
Mounting medium for histology/microscopy, Histokitt, Assistent®
Mounting medium for histology/microscopy, Histokitt, Assistent®
Catalog # HECH41025010
CAS Number:  
undefined
Mounting medium for histology/microscopy, Histokitt, Assistent®
Catalog # HECH41025010
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Histokitt mounting medium
Specifications
About this item
Mounting medium for histology/microscopy.
  • Light-resistant
  • Neutral
  • Colourless
  • Does not affect colour of the specimen
  • Soluble e.g. In benzene, chloroform, dioxane, xylene
  • In UN certified tinplate bottle
Hardening time 20 minutes, cold-resistant down to -17 °C (+1,4 °F).