Anti-CD204 (Msr1) Rat Monoclonal Antibody [clone: 1F8C33]
Catalog # BLEG154702
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:CD204 (Msr1)
- Clonality:Monoclonal
- Clone:1F8C33
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Format:Purified
- Antigen synonyms:Macrophage scavenger receptor I|SR-A|Scara1|MSR|Msr1|Scavenger receptor type A
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:Recombinant mouse CD204 extracellular domain
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography.
- Size:500 µg
- Pk:500 µG