Anti-P2RY12 Rat Monoclonal Antibody [clone: S16007D]
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG848001
- BLEG848002
- Antigen symbol:
- P2RY12
- P2RY12
- Antigen synonyms:
- P2Y12P2Y12 platelet ADP receptor|P2Y(12)R|ADPG-R|BDPLT8|G-protein coupled|P2T(AC)|P2Y12|P2Y(AC)|P2Y(ADP)|12|purinergic receptor P2Y|purinergic receptor P2RY12|HORK3|P2Y(cyc)
- P2Y12P2Y12 platelet ADP receptor|P2Y(12)R|ADPG-R|BDPLT8|G-protein coupled|P2T(AC)|P2Y12|P2Y(AC)|P2Y(ADP)|12|purinergic receptor P2Y|purinergic receptor P2RY12|HORK3|P2Y(cyc)
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- S16007D
- S16007D
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Mouse P2RY12.
- Mouse P2RY12.
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, FC
- IHC-P, FC
Specifications
Frequently Bought Together