Anti-alpha-Synuclein Mouse Monoclonal Antibody [clone: Syn303]
Catalog # BLEG824301
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:alpha-Synuclein
- Clonality:Monoclonal
- Clone:Syn303
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Format:Azide-free
- Antigen synonyms:PARK1|synuclein alpha-140|PARK4|Alpha-synuclein|NACP|PD1
- Storage buffer:Phosphate-buffered solution.
- Storage temperature:2…8 °C
- Concentration:1 mg/ml
- Tested applications:IHC-P
- Purification:The antibody was purified by affinity chromatography.
- Size:100 µl
- Pk:100 µl