Anti-NMDAR1 Mouse Monoclonal Antibody [clone: N308/48]
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG818602
- BLEG818601
- Antigen symbol:
- NMDAR1
- NMDAR1
- Antigen synonyms:
- NMDAR1 receptor C1 cassette|NMD-R1|Glutamate receptor ionotropic|glutamate receptor subunit zeta-1|NMDA 1|GluN1/NR1 glutamate receptor|N-methyl-D-aspartate glutamate receptor|N-methyl-D-aspartate receptor subunit NR1|neurotransmitter receptor
- NMDAR1 receptor C1 cassette|NMD-R1|Glutamate receptor ionotropic|glutamate receptor subunit zeta-1|NMDA 1|GluN1/NR1 glutamate receptor|N-methyl-D-aspartate glutamate receptor|N-methyl-D-aspartate receptor subunit NR1|neurotransmitter receptor
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- N308/48
- N308/48
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- This monoclonal antibody was raised against a fusion protein corresponding to amino acids 42-361 (extracellular, N-terminus) of rat GluN1/NR1 glutamate receptor protein.
- This monoclonal antibody was raised against a fusion protein corresponding to amino acids 42-361 (extracellular, N-terminus) of rat GluN1/NR1 glutamate receptor protein.
- Format:
- Azide-free
- Azide-free
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Phosphate-buffered solution.
- Phosphate-buffered solution.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- WB, IHC
- WB, IHC
Specifications
Frequently Bought Together