Anti-Tubulin beta 3 (TUBB3) Mouse Monoclonal Antibody [clone: TUJ1]
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Catalog No:
- BLEG801202
- BLEG801213
- Antigen symbol:
- Tubulin beta 3 (TUBB3)
- Tubulin beta 3 (TUBB3)
- Antigen synonyms:
- tubulin beta-4 chain|Tubulin beta-3 chain|CDCBM|CFEOM3A|tubulin beta-III|class III beta-tubulin|FEOM3|CDCBM1|TUBB4|CFEOM3
- tubulin beta-4 chain|Tubulin beta-3 chain|CDCBM|CFEOM3A|tubulin beta-III|class III beta-tubulin|FEOM3|CDCBM1|TUBB4|CFEOM3
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- TUJ1
- TUJ1
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- This antibody was raised against microtubules derived from rat brain.
- This antibody was raised against microtubules derived from rat brain.
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Phosphate-buffered solution + 0.03% Thimerosal.
- Phosphate-buffered solution + 0.03% Thimerosal.
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- IHC-P, WB, ICC, FC
- IHC-P, WB, ICC, FC
Specifications
Frequently Bought Together