J.T.Baker® BAKERBOND®, Supported Liquid Extraction Plates
Supplier: Avantor
Certificates
About this item
J.T.Baker® BAKERBOND® supported liquid extraction (SLE) plates are an ideal and faster alternative to traditional liquid-liquid extraction methods and allow excellent recoveries of analytes from biological matrices.
- Diatomaceous earth packing material
- Simple protocol
- Quicker and easier than liquid-liquid extraction
- 96-well plate or cartridge formats offered
- Can be used for protein removal
This sample preparation method can be used prior to chromatographic methods like LC-MS, HPLC and UHPLC, and has wide applicability for acids, bases and neutral compounds.
Related Information
J T Baker Protein Precipitation and Supported Liquid Extraction Plates
Notes d'application
Frequently Bought Together