ContactUsLinkComponent
PCR plates, 96-well, fully skirted, exo-frame
PCR plates, 96-well, fully skirted, exo-frame
Catalog # ISTSIST-601-096GCT
Supplier:  iST Scientific Ltd
undefined
PCR plates, 96-well, fully skirted, exo-frame
Catalog # ISTSIST-601-096GCT
Supplier:  iST Scientific Ltd
Specifications
  • Frame colour:
    Grey
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Pk:
    50
  • Well volume:
    0,2 ml
Specifications
About this item
Fully skirted 96-well PCR plate with clear PP wells and grey PC frame.
  • Free of DNA, DNase, RNase, and PCR inhibitors
  • Low binding PP wells
  • Alpha numberic labeling
  • Manufactured under the most modern clean room conditions
  • Automation friendly
Rigid two-component design eliminates warping and distortion during PCR.
Packaging: Packaged 10 plates per sleeve, 50 plates per box.