Order Entry
Belgium
ContactUsLinkComponent
Live-or-Dye™ Yeast fixable live/dead staining kit
Live-or-Dye™ Yeast fixable live/dead staining kit
Catalog # 31064.
Supplier:  Biotium
CAS Number:  
undefined
Live-or-Dye™ Yeast fixable live/dead staining kit
Catalog # 31064.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Yeast live-or-dye fixable live/dead staining kit, Green/Red
  • Colour:
    Green/Red
  • Pk Info:
    1000 assays

Specifications

About this item

The Yeast Live-or-Dye™ Fixable Live/Dead Staining Kit is a fixable, fluorescent assay to determine yeast viability. The kit contains two fluorescent components, Thiazole Orange and Live-or-Dye™ 568/583.

  • Stable and selective dead cell staining
  • Simple, rapid, no-wash staining protocol
  • Tolerates formaldehyde fixation
  • Suitable for flow cytometry and microscopy

Live-or-Dye™ 568/583 is a cell membrane impermeable amine-reactive dye. It is able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. This dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. It is suitable for detection by fluorescence microscopy or flow cytometry.

Thiazole Orange is a green fluorescent dye that localizes to the cytoplasm, with some nuclear accumulation. It is fixable, resulting in diffuse cytoplasmic staining in fixed cells.

After staining yeast with this kit, both living and dead cells will be stained with green fluorescence. Only dead cells will be stained with red fluorescence.

Spectral Properties
Live-or-Dye™ 568/583 Ex/Em 568/583 nm
Thiazole Orange Ex/Em 512/533 nm

Delivery: Kit includes Live-or-Dye™ 568/583 (solid), four vials, Thiazole Orange 10 mM in DMSO, 100 µl and Anhydrous DMSO, 250 µl