Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

SFB expansion

SFB expansion, R10.4.1 flow cells, 4, 1 KIT
Catalog # OXNTEXP-SFB001
SFB expansion
Catalog # OXNTEXP-SFB001
CAS Number:  
SFB expansion
SFB expansion

SFB expansion, R10.4.1 flow cells, 4, 1 KIT

CAS Number:  
Price
34,20 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Short Fragment Buffer (SFB).

This kit is recommended for users who require additional Short Fragment Buffer for use with the 'PCR tiling of COVID-19 virus' protocol or any other experiment sequencing short fragments.

SFB Expansion contains four vials of Short Fragment Buffer (SFB), sufficient for four reactions.

 : Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.

 : Oxford Nanopore Technologies products are currently for research use only.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    SFB expansion
  • Compatible with:
    R10.4.1 flow cells
  • No. of reactions:
    4

Specifications