Order Entry
Belgium
ContactUsLinkComponent
Sequencing auxiliary vials
Sequencing auxiliary vials
Catalog # OXNTEXP-AUX003
CAS Number:  
Sequencing auxiliary vials
Catalog # OXNTEXP-AUX003
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Sequencing auxiliary vials v14
  • No. of reactions:
    12

Specifications

About this item

Additional sequencing buffer, elution buffer, and loading beads.

These kits are recommended for users who would like to split a library across multiple flow cells, need to add more library/a new library onto a washed flow cell, or wish to top up a flow cell with additional library.

Sequencing Auxiliary Vials (EXP-AUX003) contain Sequencing Buffer (SB), Elution Buffer (EB), Library Solution (LIS) and Library Beads (LIB) with v14 flow cell priming reagents: Flow Cell Flush (FCF) and Flow Cell Tether (FCT). This product is compatible with R10.4.1 Flow Cells and all kits using Kit 14 chemistry (e.g. Ligation Sequencing Kit SQK-LSK114).

Delivery: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.