ContactUsLinkComponent
Test strips, QUANTOFIX Chlorine Sensitive 1
Test strips, QUANTOFIX Chlorine Sensitive 1
Catalog # 634-1189
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, QUANTOFIX Chlorine Sensitive 1
Catalog # 634-1189
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Pk:
    50
  • Description:
    QUANTOFIX Chlorine Sensitive 1, 50 tests
Specifications
About this item
Semi-quantitative test strips for reliable determination of chlorine concentrations as low as 0,05 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity by a factor of nearly 10. Ideal for rapid and easy analysis of chlorine directly on-site. No maintenance or calibration is required.
  • Sensitive rapid test