ContactUsLinkComponent
QUANTOFIX Iron Sensitive 1
QUANTOFIX Iron Sensitive 1
N° de catalogue MANA91359
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
QUANTOFIX Iron Sensitive 1
N° de catalogue MANA91359
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Pk:
    25
  • Description:
    QUANTOFIX Iron Sensitive 1, 25 tests
Specifications
About this item
Semi-quantitative test strips for reliable determination of iron concentrations as low as 0.05 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity by a factor of nearly 10. Ideal for rapid and easy analysis of iron directly on-site. No maintenance or calibration is required.
  • Sensitive rapid test