Order Entry
Belgium
ContactUsLinkComponent
HPLC columns, PFAS Delay, NUCLEODUR®
HPLC columns, PFAS Delay, NUCLEODUR®
Catalogus # MANA760673.20
Leverancier:  MACHEREY-NAGEL
HPLC columns, PFAS Delay, NUCLEODUR®
Catalogus # MANA760673.20
Leverancier:  MACHEREY-NAGEL

Specificaties

  • Column Format:
    Analytical
  • Ø int.:
    2,00 mm
  • Particle Size:
    5 µm
  • Pore size:
    110 Å
  • Max. pressure:
    600 bar
  • Length:
    50 mm
  • Shipping conditions:
    Eluent in HPLC column
    Acetonitrile – water
  • Endcapped:
    Yes
  • pH range:
    1,0 - 9,0
  • Surface area:
    340 m²/g
  • Temperature limit:
    60 °C
    40 °C (phosphate buffer)
  • Application:
    PFAS compounds
  • Storage:
    RT
  • Column hardware:
    Stainless steel
  • Pk:
    1

Specificaties

Over dit artikel

NUCLEODUR PFAS Delay columns trap and therefor delay system-related PFAS contaminants, ensuring trace level analysis of PFAS in your samples. NUCLEODUR PFAS Delay reversed phase is hydrophobic with distinct polar selectivity for special PFAS analysis. Suitability for PFAS analysis is assured through qualified batch analysis.

  • High purity spherical silica gel
  • Polar selectivity
  • Special PFAS analysis
  • Low bleeding characteristics

This HPLC phase is made of high purity NUCLEODUR HPLC silica gel and is extensively endcapped. Due to a low bleeding characteristics these columns are suitable for high sensitivity LC/MS analysis.

Levering: Supplied with a test chromatogram, instruction leaflet and batch test certificate.