Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

HiLoad Superdex columns, 200 PG

Supplier:   Cytiva
HiLoad Superdex columns, 200 PG
Supplier:  Cytiva
Certificates
HiLoad Superdex columns, 200 PG

2 Options Available Below

Product Details & Documents

Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component

  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • 1/16" fittings for convenient connections to ÄKTA design systems

Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex 30 pg), Mr 3 000 to 70 000 (Superdex 75 pg), and Mr 10 000 600 000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.

 : Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.

Specifications for Product Family

Specifications

  • Catalog No:
  • 28-9893-35
  • 28-9893-36
  • Max. pressure:
  • 3 bar
  • 3 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Product Family Options

Product Information

  • Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Sample volume
    Flow rate
    Flow velocity
    Availability
    Price
  • 28-9893-35
    HiLoad™ 16/600 Superdex™ 200 PG
    HiLoad™ 16/600 Superdex™ 200 PG
    Glass
    34 µm
    16×600 mm
    120 ml
    ≤ 5 ml
    1,6 ml/min
    10 - 50 cm/h
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 930,00 €
    2 930,00 €
    1 * 1 ST
       2 930,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    28-9893-35
    Max. pressure
    3 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol
  • 28-9893-36
    HiLoad™ 26/600 Superdex™ 200 PG
    HiLoad™ 26/600 Superdex™ 200 PG
    Glass
    34 µm
    26×600 mm
    320 ml
    ≤ 13 ml
    1,6 ml/min
    10 - 50 cm/h
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 400,00 €
    4 400,00 €
    1 * 1 ST
       4 400,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    28-9893-36
    Max. pressure
    3 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol