ContactUsLinkComponent
Syringe filters, PP, Puradisc™, Whatman™
Syringe filters, PP, Puradisc™, Whatman™
Certificates
undefined
Syringe filters, PP, Puradisc™, Whatman™
Certificates
Specifications
  • Catalog No:
  • 514-0237
  • 514-8030
  • 516-1434
  • 514-8032
  • Ø:
  • 25 mm
  • 25 mm
  • 25 mm
  • 13 mm
  • Dimensions:
  • 22,9×28,4 mm
  • 22,9×28,4 mm
  • 22,9×28,4 mm
  • 16,3×19,8 mm
  • Max. operating pressure:
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • 5,2 bar
  • Weight:
  • 2,7 g
  • 2,7 g
  • 2,7 g
  • 0,95 g