Quick-lock connectors
About this item
Easy and safe connection of capillary tubing and SCAT Safety Caps using quick couplings. Connect and disconnect with a "click" - without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
- Made of chemically resistant glass-filled polypropylene, PEEK® materials
- Leak protection thanks to integrated shut-off valves
- Closed system for drip-free disconnection with very low air entrapment
- Minimizes contamination and no need to purge air bubbles from the system
- Suitable for capillaries with outer diameter 1,6/2,3/3,2 mm
- User-friendly, ergonomic design, enables safe connection
- Up to vacuum 15 psi, 1,0 bar pressure resistance
The quick-lock connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
5 Options Available Below
Product Details & Documents
Easy and safe connection of capillary tubing and SCAT Safety Caps using quick couplings. Connect and disconnect with a "click" - without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
- Made of chemically resistant glass-filled polypropylene, PEEK® materials
- Leak protection thanks to integrated shut-off valves
- Closed system for drip-free disconnection with very low air entrapment
- Minimizes contamination and no need to purge air bubbles from the system
- Suitable for capillaries with outer diameter 1,6/2,3/3,2 mm
- User-friendly, ergonomic design, enables safe connection
- Up to vacuum 15 psi, 1,0 bar pressure resistance
The quick-lock connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
Product Family Options
Single quick-lock connectors
Product Information
- ForAvailabilityPrice
- SCTV360179Quick-lock connector (f), includes 3× PFA-fitting (1,6/2,3/3,2 mm ID)Quick-lock connector (f), includes 3× PFA-fitting (1,6/2,3/3,2 mm ID)1 capillary tubeLog in for availability1 * 1 ST37,60 €
- SCTV360180Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm ID)Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm ID)1 Safety CapLog in for availability1 * 1 ST37,60 €
- SCTV360190Quick-lock connector (m), includes 3× PFA-fitting (1,6/2,3/3,2 mm ID)Quick-lock connector (m), includes 3× PFA-fitting (1,6/2,3/3,2 mm ID)1 capillary tubeLog in for availability1 * 1 ST37,60 €
Quick-lock connector sets
Product Information
- ForAvailabilityPrice
- SCTV360183Quick-lock connector Set I, includes SCTV:360179 Quick Couplings (f) and SCTV360180 Quick coupling (m)Quick-lock connector Set I, includes SCTV:360179 Quick Couplings (f) and SCTV360180 Quick coupling (m)1 capillary tube and 1 Safety CapLog in for availability1 * 1 ST75,30 €
- SCTV360189Quick-lock connector Set II, includes SCTV360179 Quick Coupling (f) and SCTV360190 Quick coupling (m)Quick-lock connector Set II, includes SCTV360179 Quick Coupling (f) and SCTV360190 Quick coupling (m)2 capillary tubesLog in for availability1 * 1 ST75,30 €