VWR®, LED-Fluorescence Cubes
For use with microscope series TL534B FL.
For use with microscope series TL534B FL.
Product Family Options
- DescriptionOptical features
- Blue LED fluorescence cube for TL534B FLLED emission 460 nm,
Excitation filter 455-495 nm,
Dichroic mirror cut-off 500 nm,
Emission filter 510 LP nm1 * 1 ST968,00 € - Green LED fluorescence cube for TL534B FLLED emission 523 nm,
Excitation filter 510-550 nm,
Dichroic mirror cut-off 570 nm,
Emission filter 575 LP nm1 * 1 ST1 160,00 € - UV LED fluorescence cube for TL534B FLLED emission 365 nm,
Ecitation filter 325 - 375 nm,
Dichroic mirror cut-off 415 nm,
Emission filter 435 LP nm1 * 1 ST1 690,00 € - Red 1 LED fluorescence cube for TL534B FLLED emission 623 nm,
Excitation filter 590 - 650 nm,
Dichroic mirror cut-off 660 nm,
Emission filter 665 LP nm1 * 1 ST3 920,00 € - Red 2 LED fluorescence cube for TL534B FLLED emission 623 nm,
Ecitation filter 595-645 nm,
Dichroic mirror cut-off 655 nm,
Emission filter 665 - 715 nm1 * 1 ST3 920,00 € - Deep red LED fluorescence cube for TL534B FLLED emission 660 nm,
Excitation filter 623 - 678 nm,
Dichroic mirror cut-off 685 nm,
Emission filter 690 - 750 nm1 * 1 ST3 920,00 € - Far red LED fluorescence cube for TL534B FLLED emission 740 nm,
Excitation filter 720 - 760 nm,
Dichroic mirror cut-off 770 nm,
Emission filter 780 LP nm1 * 1 ST3 930,00 € - Amber LED fluorescence cube for TL534B FLLED emission 590 nm,
Excitation filter 582 - 603 nm,
Dichroic mirror cut-off 610 nm,
Emission filter 615 - 645 nm1 * 1 ST3 940,00 € - Violet LED fluorescence cube for TL534B FLLED emission 405 nm,
Excitation filter 390 - 420 nm,
Dichroic mirror cut-off 440 nm,
Emission filter 450 LP nm1 * 1 ST1 670,00 €



