Orders LinkContactUsLinkComponent
VWR®, L-Glutamine
VWR®, L-Glutamine
L-Glutamine, 100X, 200 mM, Liquid, 100 mL
Catalog # 392-0441
Supplier:   VWR Chemicals
VWR®, L-Glutamine
Catalog # 392-0441
Supplier:  VWR Chemicals
CAS Number:  
VWR®, L-Glutamine
VWR®, L-Glutamine
L-Glutamine, 100X, 200 mM, Liquid, 100 mL
CAS Number:  
Price
9,90 €
Log in for availability
1 * 100 mL
Product Details & Documents
L-Glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture.
Specifications
Specifications
  • Media Format:
    Liquid
  • Pk:
    100 mL
  • Description:
    L-Glutamine, 100X, 200 mM
Specifications