Orders LinkContactUsLinkComponent
High efficiency GC columns
High efficiency GC columns
Supplier:   VARIAN
High efficiency GC columns
Supplier:  VARIAN
Certificates
High efficiency GC columns
High efficiency GC columns
8 Options Available Below
Product Details & Documents
High efficiency 0,15 and 0,18 mm Int.Ø columns are a simple and cost effective way to increase sample throughput with existing GC or GC/MS instruments.
  • Increase sample throughput and productivity while maintaining resolution
  • Reduce carrier gas usage and cost per analysis
  • Operates with He or H₂ carrier gas
  • Available in more than 20 stationary phases
This leading-edge column technology is ideal for applications that require faster run times, such as high-throughput screening, fast process monitoring, and fast method development. Agilent High Efficiency GC columns can reduce sample run time by 50% or more without compromising resolution.
Product Family Options
Product Information
  • Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • CP-Sil 13 CB
    0,63 µm
    0,15 mm
    12 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 470,00 €
    2 470,00 €
    1 * 1 ST
       2 470,00 €
  • CP-Sil 8 CB low bleed/ms
    0,15 µm
    0,15 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 613,00 €
    613,00 €
    1 * 1 ST
       613,00 €
  • CP-Sil 5 CB
    0,15 µm
    0,15 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 524,00 €
    524,00 €
    1 * 1 ST
       524,00 €
  • CP-Sil 5 CB low bleed/ms
    0,15 µm
    0,15 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 613,00 €
    613,00 €
    1 * 1 ST
       613,00 €
  • VF-1ms
    0,60 µm
    0,15 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 877,00 €
    877,00 €
    1 * 1 ST
       877,00 €
  • VF-1ms
    1,00 µm
    0,15 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 250,00 €
    1 250,00 €
    1 * 1 ST
       1 250,00 €
  • VF-WAXms
    0,25 µm
    0,15 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 890,00 €
    890,00 €
    1 * 1 ST
       890,00 €
  • VF-1ms
    0,60 µm
    0,15 mm
    40 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €