Orders LinkContactUsLinkComponent
Columns, PlusPore
Columns, PlusPore
Supplier:   Agilent
Columns, PlusPore
Supplier:  Agilent
Certificates
Columns, PlusPore
4 Options Available Below
Product Details & Documents
The PlusPore family offers a major resolution improvement over mixed medias and wider range than single pore columns.
  • Separation is maximised over four common MW ranges
  • Small particles and large pore volumes give very fast, high resolution
  • High speed chromatography offers 6 min separations for common samples
The presence of multiple pore sizes on each particle maximises the separation efficiency while still offering a wide MW range with no dislocations.
Product Family Options
Product Information
  • Column Format
    Ø int.
    Length
    Availability
    Price
  • Analytical
    2,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 680,00 €
    2 680,00 €
    1 * 1 ST
       2 680,00 €
  • Analytical
    2,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 680,00 €
    2 680,00 €
    1 * 1 ST
       2 680,00 €
  • Analytical
    2,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 370,00 €
    2 370,00 €
    1 * 1 ST
       2 370,00 €
  • Analytical
    2,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 630,00 €
    2 630,00 €
    1 * 1 ST
       2 630,00 €