GC columns, J&W CAM
: Agilent
CAM is a base-deactivated PEG column specifically designed for amine analysis.
- Excellent peak shape for primary animes
- Replaces HP-Basicwax
- Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
The column is not bonded or cross-linked, and so we do not recommend solvent rinsing.
Frequently Bought Together