Orders LinkContactUsLinkComponent
Stopcocks, with GL screw thread
Stopcocks, with GL screw thread
Supplier:   Bohlender
Stopcocks, with GL screw thread
Supplier:  Bohlender
Certificates
Stopcocks, with GL screw thread
Stopcocks, with GL screw thread
9 Options Available Below
Product Details & Documents
PTFE.
  • Excellent chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −30 to +150 °C
  • Pressure resistant up to a maximum of 6 bar, vacuum suitable
Two-way stopcocks with straight bore and two threaded connectors or three-way stopcocks with L-shaped or T-shaped bore with three threaded connectors. Cylindrical stopcock for good leakproof properties, stop valve with flow direction marking. To distribute liquids or gases. Fast and simple interruption of the flow. Connection of hoses or tubes using GL laboratory screw connections.
Product Family Options
2-way stopcocks
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €
  • 8 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 203,00 €
    203,00 €
    1 * 1 ST
       203,00 €
3-way stopcocks, L-shaped bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
  • 6 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 209,00 €
    209,00 €
    1 * 1 ST
       209,00 €
  • 6 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
3-way stopcocks, T-shaped bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 6 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 300,00 €
    300,00 €
    1 * 1 ST
       300,00 €
  • 4 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 270,00 €
    270,00 €
    1 * 1 ST
       270,00 €
  • 4 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €