Order Entry
Belgium
ContactUsLinkComponent
VWR®, Certified Reference Materials for the standardisation of volumetric solutions
VWR®, Certified Reference Materials for the standardisation of volumetric solutions
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Certified Reference Materials for the standardisation of volumetric solutions
Supplier:  VWR Chemicals
Certificates

Specifications

  • CAS No.:
    7440-66-6
  • Formula:
    Zn
  • Description:
    Zinc
  • MDL No.:
    MFCD00149220

Specifications

Frequently Bought Together