Cycloheximide 100 mM
1 mL
Catalog # API-03
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).
- Supplied as 1ml 100mM solution.
G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.
Cycloheximide is an inhibitor of protein synthesis by interfering with the translocation step (movement of two tRNA molecules and mRNA on ribosome) in protein synthesis, thus blocking translational elongation.
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35 g/mol
Specifications
Specifications
- Pk:1 mL



