ContactUsLinkComponent
Murashige and skoog modified basal salts (½ Micros and ½ Macros)
Murashige and skoog modified basal salts (½ Micros and ½ Macros)
Catalog # APOSPMM153-50L
Supplier:  Apollo Scientific
CAS Number:  
Murashige and skoog modified basal salts (½ Micros and ½ Macros)
Catalog # APOSPMM153-50L
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    50 L
  • Description:
    Murashige and Skoog modified basal salts (½ micros and ½ macros) 50 L
  • Seq:
    03
Specifications
About this item
Contains macro/micronutrients, sucrose, agar and organics as described by Murashige and Skoog, 1962 Plant tissue culture testedFull specification given in appendix.
  • MFCD00240976