Heller's basal salt mixture
Contains macro/micronutrients as described by Heller, 1953 Plant tissue culture testedFull specification given in appendix.
- MFCD00283413
Frequently Bought Together
Contains macro/micronutrients as described by Heller, 1953 Plant tissue culture testedFull specification given in appendix.