ContactUsLinkComponent
Stabiliser for proteins and standards, SeramunStab® Stpur
Stabiliser for proteins and standards, SeramunStab® Stpur
Catalog # APOSBIL3118-100ML
Supplier:  Apollo Scientific
CAS Number:  
Stabiliser for proteins and standards, SeramunStab® Stpur
Catalog # APOSBIL3118-100ML
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    SeramunStab® Stpur stabiliser for proteins and standards
  • Seq:
    01
Specifications
About this item
Liquid product colourless to slightly yellow or greenish odourless pH 7,3 to 7,5 boiling point 104 °C. Stabiliser for controls and protein standards.