HPLC columns, ZORBAX® Eclipse, XDB-C8
À propos de cet article
Agilent ZORBAX® Eclipse XDB-C8 columns provide four bonded phase choices for method development optimisation.
- Long lifetime
- Four selectivity choices for method development optimisation
- Good peak shape for basic, acidic and neutral compounds
- High performance over a wide pH range pH 2 to 9
- Particle sizes available in 1,8, 3,5, 5 and 7 μm for high speed, high resolution, analytical and preparative scale separations
These columns provide good peak shape over a wide pH range (2 to 9) for additional method development flexibility with one family of columns. Eclipse XDB columns can be used for method development at low pH (2 to 3) and the same column can be used for method development in the mid pH (6 to 8) region. In the mid pH region, residual silanols are more active and tailing interactions are more likely. To overcome these interactions, Eclipse XDB columns are extra densely bonded and double end capped through a proprietary process to cover as many active silanols as possible. The result is superior peak shape of basic compounds from pH 2 to 9.
37 Options disponibles ci-dessous
Product Details & Documents
Agilent ZORBAX® Eclipse XDB-C8 columns provide four bonded phase choices for method development optimisation.
- Long lifetime
- Four selectivity choices for method development optimisation
- Good peak shape for basic, acidic and neutral compounds
- High performance over a wide pH range pH 2 to 9
- Particle sizes available in 1,8, 3,5, 5 and 7 μm for high speed, high resolution, analytical and preparative scale separations
These columns provide good peak shape over a wide pH range (2 to 9) for additional method development flexibility with one family of columns. Eclipse XDB columns can be used for method development at low pH (2 to 3) and the same column can be used for method development in the mid pH (6 to 8) region. In the mid pH region, residual silanols are more active and tailing interactions are more likely. To overcome these interactions, Eclipse XDB columns are extra densely bonded and double end capped through a proprietary process to cover as many active silanols as possible. The result is superior peak shape of basic compounds from pH 2 to 9.
Product Family Options
Analytical columns
Informations sur le produit
- Column FormatPhaseParticle SizeØ int.LengthAvailabilityPrice
Semi micro and micro column
Informations sur le produit
- Column FormatPhaseParticle SizeØ int.LengthAvailabilityPrice
Guard cartridge
Informations sur le produit
Method validation kit
Informations sur le produit
- Column FormatPhaseParticle SizeØ int.LengthAvailabilityPrice
- Method Validation KitEclipse XDB-C81,8 µm2,10 mm50 mmConnexion Pour la disponibilité1 * 1 ST2 440,00 €
- Method Validation KitEclipse XDB-C81,8 µm2,10 mm100 mmConnexion Pour la disponibilité1 * 1 ST2 820,00 €
- Method Validation KitEclipse XDB-C81,8 µm3,00 mm50 mmConnexion Pour la disponibilité1 * 1 ST2 440,00 €
- Method Validation KitEclipse XDB-C81,8 µm3,00 mm100 mmConnexion Pour la disponibilité1 * 1 ST2 820,00 €
- Method Validation KitEclipse XDB-C81,8 µm4,60 mm50 mmConnexion Pour la disponibilité1 * 1 ST2 420,00 €
- Method Validation KitEclipse XDB-C83,5 µm4,60 mm50 mmConnexion Pour la disponibilité1 * 1 ST1 740,00 €
- Method Validation KitEclipse XDB-C81,8 µm4,60 mm100 mmConnexion Pour la disponibilité1 * 1 ST2 820,00 €
- Method Validation KitEclipse XDB-C83,5 µm4,60 mm100 mmConnexion Pour la disponibilité1 * 1 ST2 320,00 €
- Method Validation KitEclipse XDB-C83,5 µm4,60 mm150 mmConnexion Pour la disponibilité1 * 1 ST2 720,00 €
- Method Validation KitEclipse XDB-C85 µm4,60 mm150 mmConnexion Pour la disponibilité1 * 1 ST2 420,00 €
- Method Validation KitEclipse XDB-C85 µm4,60 mm250 mmConnexion Pour la disponibilité1 * 1 ST2 640,00 €