Order Entry
Belgium
ContactUsLinkComponent
Brilliant probe multiplex MM
Brilliant probe multiplex MM
Catalog # AGLS600553
Supplier:  AGILENT
CAS Number:  
Brilliant probe multiplex MM
Catalog # AGLS600553
Supplier:  AGILENT
CAS Number:  

Specifications

  • Pk:
    1
  • Application:
    qPCR probe
  • Multiplexing:
    Yes
  • Description:
    Brilliant multiplex QPCR master mix, no.of.reactions: 200 (25 µl/reaction)

Specifications

About this item

The brilliant multiplex QPCR master mix allows you to amplify up to four targets in a single real-time PCR Reaction.

  • Maximises analysis of limited or rare samples
  • Allows detection of multiple targets plus an internal control
  • More economical per sample than singleplex, while saving time and increasing throughput

It provides sufficient reaction components to accurately quantify both low-and-high-abundance targets in the same tube.

The sensitivity remains equivalent to that seen in singleplex.