
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% (by TLC)
Catalog # AGLS300127
Supplier: AGILENT
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity:≥99% (TLC)
Specification Test Results
About this item
:
:238.31 g/mol
Melting Pt:112...114 °C
:GHS07
Storage Temperature:Freezer
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273