Order Entry
Belgium
Orders LinkContactUsLinkComponent
DNase I

DNase I

Supplier:   AGILENT
DNase I
Supplier:  AGILENT
Certificates

2 Options Available Below

Product Details & Documents

Purified DNase I tested to be free of contaminating activity for the degradation of nucleic acids.

  • RNase free
  • Contains a mixture of four isoenzymes from bovine pancreas (10 units/µl)

DNase I, RNase Free catalyses the degradation of double-stranded DNA into oligonucleotides 1,2 and mononucleotides. This enzyme has been isolated as a mixture of four isoenzymes from bovine pancreas that cut preferentially next to pyrimidine nucleotides.

Reaction Conditions: 40 mM Tris-HCl (pH 7,5), 6 mM MgCl₂, 2 mM CaCl₂, 1 µg DNA and 1 unit enzyme in a 25 µl volume. Incubate 30 minutes at 37 °C.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • AGLS600031
  • AGLS600032
  • Enzyme name:
  • DNase I
  • DNase I
  • Form:
  • Premixed
  • Premixed
  • Formulation:
  • 10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)
  • 10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)

Specifications

Product Family Options

Product Information

  • Pk Info
    Pk
    Availability
    Price
  • 1000 units
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 205,00 €
    205,00 €
    1 * 1 ST
       205,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    AGLS600031
    Enzyme name
    DNase I
    Form
    Premixed
    Formulation
    10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)
  • 5000 units
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 411,00 €
    411,00 €
    1 * 1 ST
       411,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    AGLS600032
    Enzyme name
    DNase I
    Form
    Premixed
    Formulation
    10 mM Tris-HCl (pH 7,5), 10 mM CaCl₂, 10 mM MgCl₂ and 50 % glycerol (v/v). (Dilutions should be made in this buffer.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.