Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
Supplier:   VARIAN
Spray chambers and brackets
Supplier:  VARIAN
Certificates
Spray chambers and brackets
Spray chambers and brackets
10 Options Available Below
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Product Family Options
Product Information
  • Availability
    Price
  • HEWL3710035300
    Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
    Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL3710037800
    Replacement tubing for ketone based organic samples, per meter
    Replacement tubing for ketone based organic samples, per meter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,00 €
    34,00 €
    1 * 1 ST
       34,00 €
  • HEWL6910006000
    O-ring, nitrile, ¹¹/₁₆" int. Ø, ¹³/₁₆" ext. Ø, ¹/₁₆" thick, for Mark VI and Sturman-Masters spray chambers for AA/ICP-OES
    O-ring, nitrile, ¹¹/₁₆" int. Ø, ¹³/₁₆" ext. Ø, ¹/₁₆" thick, for Mark VI and Sturman-Masters spray chambers for AA/ICP-OES
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €
  • HEWLG8010-60256
    Double-pass spray chamber, glass cyclonic design with ball joint socket and UniFit drain outlet, for use with organic solvents, high matrix or high TDS samples with 5100 ICP-OES
    Double-pass spray chamber, glass cyclonic design with ball joint socket and UniFit drain outlet, for use with organic solvents, high matrix or high TDS samples with 5100 ICP-OES
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 180,00 €
    1 180,00 €
    1 * 1 ST
       1 180,00 €
  • HEWLG8010-60271
    Spray chamber, single-pass, for 5100 ICP-OES, glass cyclonic design with ball joint socket and UniFit drain outlet, for aqueous and low matrix samples where best sensitivity and lowest LODs are required
    Spray chamber, single-pass, for 5100 ICP-OES, glass cyclonic design with ball joint socket and UniFit drain outlet, for aqueous and low matrix samples where best sensitivity and lowest LODs are required
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 874,00 €
    874,00 €
    1 * 1 ST
       874,00 €
  • HEWLG8010-68011
    PuraFlex transfer tube for IsoMist used with 5100 ICP-OES, required to connect aerosol outlet from IsoMist temperature-controlled spray chamber to easy-fit torch
    PuraFlex transfer tube for IsoMist used with 5100 ICP-OES, required to connect aerosol outlet from IsoMist temperature-controlled spray chamber to easy-fit torch
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 495,00 €
    495,00 €
    1 * 1 ST
       495,00 €
  • HEWLG8010-80036
    UniFit drain connector, 2,0 mm ext. Ø, 0,86 mm int. Ø, for spray chamber used with 5100 dual view ICP-OES, with 480 mm capillary tubing
    UniFit drain connector, 2,0 mm ext. Ø, 0,86 mm int. Ø, for spray chamber used with 5100 dual view ICP-OES, with 480 mm capillary tubing
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 108,00 €
    108,00 €
    1 * 3 ST
       108,00 €
  • HEWLG8010-80042
    Replacement helix seal for ICP spray chamber, for any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
    Replacement helix seal for ICP spray chamber, for any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 136,00 €
    136,00 €
    1 * 4 ST
       136,00 €
  • HEWLG8010-80043
    Replacement helix locking screw for ICP spray chamber, suits any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
    Replacement helix locking screw for ICP spray chamber, suits any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,00 €
    96,00 €
    1 * 1 ST
       96,00 €
  • VARI110868000
    Bracket for mounting multi-mode sample introduction system (MSIS) on axial and radial ICP, for all 700 series
    Bracket for mounting multi-mode sample introduction system (MSIS) on axial and radial ICP, for all 700 series
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €