Orders LinkContactUsLinkComponent
Serological pipettes
Serological pipettes
Supplier:   GILSON
Serological pipettes
Supplier:  GILSON
Certificates
Serological pipettes
Serological pipettes
6 Options Available Below
Product Details & Documents
The Gilson disposable plastic serological pipettes are perfect for use with MACROMAN® to help you in your daily cell-culture, molecular biology and microbiology applications. Ideal for scientists who need to easily and quickly transfer liquid aliquots from one container to another with precision.
  • USP Class VI polystyrene and are free of contaminants that can alter your experiment
  • Gamma-sterilised, non-pyrogenic, and certified free of detectable RNase/DNase
  • Polyester fiber plug, offering a high level of protection against over-pipetting and contamination by liquids and aerosols
  • Colour-coded rings for easy size identification
  • Dark ascending and descending graduations so you can easily read the aspirated and dispensed volumes, they also have negative graduations for extra capacity
Available in models ranging from 1 to 50 ml, these serological pipettes are reco mmended for use with MACROMAN® but can also be used with any pipette controller. They enable precise and convenient dispensing of different kinds of liquids for a variety of tasks. Two kinds of packaging for various applications and precise sampling.
Product Family Options
Product Information
  • Capacity
    Packed
    Availability
    Price
  • 1 ml
    Individually wrapped, 1000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 120,00 €
    120,00 €
    1 * 1 ST
       120,00 €
  • 2 ml
    Individually wrapped, 1000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 120,00 €
    120,00 €
    1 * 1.000 ST
       120,00 €
  • 5 ml
    Individually wrapped, 200
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 46,00 €
    46,00 €
    1 * 200 ST
       46,00 €
  • 10 ml
    Bulk, 20 bags of 25
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
  • 25 ml
    Bulk, 10 bags of 25
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST
  • 50 ml
    Individually wrapped, 100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 98,70 €
    98,70 €
    1 * 100 ST
       98,70 €