Orders LinkContactUsLinkComponent
Square Buckets
Square Buckets
Supplier:   LAMAPLAST
Square Buckets
Supplier:  LAMAPLAST
Certificates
Square Buckets
2 Options Available Below
Product Details & Documents
Robust PP bucket with ergonomic handle.
  • Square shape
  • Flat bottom ensures total emptying and easy cleaning
  • Food contact material approved
Hot filling possible up to 95 °C.
Product Family Options
Product Information
  • Colour
    Capacity
    Dimensions
    Availability
    Price
  • LAMA1147
    Square bucket with metal handle
    Square bucket with metal handle
    White
    15 L
    280×250×345 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €
  • LAMA1137
    Square bucket with metal handle
    Square bucket with metal handle
    White
    20 L
    320×315×320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,80 €
    63,80 €
    1 * 1 ST
       63,80 €