Hermetic Tamper Evident Buckets, Rectangular
Supplier: LAMAPLAST
Jump to:
About this item
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
- Rectangular shape
- Flat bottom ensures total emptying and easy cleaning
- Especially suitable in cosmetic, pharma, paint or dye industries
- Food contact material approved
Hot filling possible up to 95 °C.
3 Options Available Below
Product Details & Documents
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
- Rectangular shape
- Flat bottom ensures total emptying and easy cleaning
- Especially suitable in cosmetic, pharma, paint or dye industries
- Food contact material approved
Hot filling possible up to 95 °C.
Documents
Product Family Options
Product Information
- ColourCapacityHeightAvailabilityPrice
- LAMA1105Rectangular bucket with plastic handle, 243×168 mm (top), 218×143 mm (base)Rectangular bucket with plastic handle, 243×168 mm (top), 218×143 mm (base)White3 L119 mmLog in for availability1 * 1 ST7,00 €
- LAMA1107Rectangular bucket with plastic handle, 286×198 mm (top), 255×168 mm (base)Rectangular bucket with plastic handle, 286×198 mm (top), 255×168 mm (base)White5 L149 mmLog in for availability1 * 1 ST8,90 €
- LAMA1109Rectangular bucket with plastic handle, 365×247 mm (top), 332×210 mm (base)Rectangular bucket with plastic handle, 365×247 mm (top), 332×210 mm (base)White11 L191 mmLog in for availability1 * 1 ST15,90 €