ContactUsLinkComponent
J.T.Baker® Plate, Dilution Strips SW
J.T.Baker® Plate, Dilution Strips SW
Catalog # 43001-0031
Supplier:  Avantor
   Low Manufacturing Impact
undefined
J.T.Baker® Plate, Dilution Strips SW
Catalog # 43001-0031
Supplier:  Avantor
   Low Manufacturing Impact
Specifications
  • Sterile:
    -
  • Colour:
    Transparent
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Pk:
    250
Specifications
About this item
Suitable for use on Dynex DSX and DS2 instruments. Deep well reservoir with eight wells.
  • Strip of 8 wells
  • Made of PP