J.T.Baker®, Plate Medio
Supplier: Avantor
Certificates
About this item
PP, 96-well storage plate with conical wells.
- Due to the tangency of J.T.Baker Plate Medio plates, less than 50% stacking space is needed
- Alphanumeric code to assist identification and registration of single samples
- Higher well rims allow an airtight seal
- Conical wells aid sample mixing
J.T.Baker Plate Medio is perfectly made for limited storage areas (fridge).
Specifications
- Catalog No:
- 43001-0101
- 43001-0103
- 43001-0102
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)