PLgel MIXED-LS column
Product Details & Documents
Eliminates particle leakage to improve data quality with light scattering detection. Typical applications: polyethylenes, polyolefins. Compatible with organic solvents.
- Obtain an instant improvement in data quality
- No need for conditioning, saving time and solvent costs
- Maximize the performance of light scattering detectors with minimal investment
The PLgel MIXED-LS series is a PS/DVB packing using an innovative polymerisation technique to virtually eliminate the nano-particle leakage from large pore gels that interferes with light scattering measurement. An immediate improvement in data quality and lower limit of detection is achieved with PLgel MIXED-LS columns in place of conventional GPC columns.
Specifications
- Column Format:Guard column
- Ø int.:7,50 mm
- Particle Size:10 µm
- Length:50 mm
- Phase:PLgel MIXED
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



