ContactUsLinkComponent
J.T.Baker®, Robotic Tips Type MP
J.T.Baker®, Robotic Tips Type MP
Catalog # 49209-0105
Supplier:  Avantor
   Low Manufacturing Impact
undefined
J.T.Baker®, Robotic Tips Type MP
Catalog # 49209-0105
Supplier:  Avantor
   Low Manufacturing Impact
Specifications
  • Packed:
    8× 5 racks, 96 each, blistered
  • Sterile:
    -
  • Volume:
    1000 µl
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Tip colour:
    Black
  • Filter:
    +
  • Pk:
    3.840
Specifications
About this item
Suitable for use with Roche MagNA Pure instruments. Available in conductive version, with filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Barcoded trays for automated identification
  • Free of DNA, DNase, RNase and PCR inhibitors
  • Ideal for high throughput analysis
  • Packed in blisters
The validation of the tips on the instrument for specific applications is within the responsibility of the user.