Orders LinkContactUsLinkComponent
Human/mouse reference RNA
Human/mouse reference RNA
Supplier:   AGILENT
Human/mouse reference RNA
Supplier:  AGILENT
Certificates
Human/mouse reference RNA
2 Options Available Below
Product Details & Documents
Total RNA which has been extracted from cell lines representing different human tissues, pooled (10 human) or pooled (11 mouse) and treated to remove DNA to undetectable levels.
  • Greater consistency than RNA references derived from tissue sources
  • Maximum representation of low, medium, and high abundant gene transcripts
  • Manufactured at industrial scale lot sizes and stringently quality controlled for minimised lot-to-lot variation
  • No detectable levels of genomic DNA
Typical uses include optimisation of primer/probe sets to ensure no cross reactivity, as a standard to relatively quantitate unknowns to an unchanging reference for immediate validation without wasting precious RNA, or to compare QPCR efficiencies across labs/platforms/experiments.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Pk Info
    Pk
    Availability
    Price
  • AGLS750500
    Quantitative PCR human reference total RNA, pool of 10 human cell lines providing broad gene coverage qualified for use in QPCR
    Quantitative PCR human reference total RNA, pool of 10 human cell lines providing broad gene coverage qualified for use in QPCR
    25 µg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 307,00 €
    307,00 €
    1 * 1 ST
       307,00 €
  • AGLS750600
    Quantitative PCR mouse reference total RNA, pool of 11 mouse cell lines providing broad gene coverage qualified for use in QPCR
    Quantitative PCR mouse reference total RNA, pool of 11 mouse cell lines providing broad gene coverage qualified for use in QPCR
    25 µg
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 227,00 €
    227,00 €
    1 * 1 ST
       227,00 €