Skip to Header
Skip to Main Content
Skip to Footer
Avancé
Avancé
Fiches de données de sécurité (FDS)
Certificats
Données du fabricant
0
0,00 €
Order Entry
Login
/
S'inscrire
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium formate
Sodium formate solution
Sodium formate solution
Sodium formate solution
Catalog # 51197-100ML
Supplier:
Merck
CAS Number:
Expand image
Sodium formate solution
Catalog # 51197-100ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
:
:
68.01 g/mol
CAS Number:
141-53-7
Formula:
HCO₂Na
MDL Number:
MFCD00013101
Product Details