Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol solution 1 M

D-(-)-Sorbitol solution 1 M

100 mL
Catalog # R038
Supplier:   G-Biosciences
D-(-)-Sorbitol solution 1 M
Catalog # R038
Supplier:  G-Biosciences
CAS Number:  50-70-4
D-(-)-Sorbitol solution 1 M
D-(-)-Sorbitol solution 1 M

100 mL

CAS Number:  50-70-4
Price
45,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

Quality DNase and RNase free reagent solution for yeast research.

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.