Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride solution 5 M, sterile
Sodium chloride solution 5 M, sterile
Catalog # R006
Supplier:  G-Biosciences
CAS Number:  
Sodium chloride solution 5 M, sterile
Catalog # R006
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477